Corning Disposable Vacuum Filtration Systems
Ideal for filtering cell culture media, biological fluids, and aqueous solutions
  • Large membrane diameters (50, 70, and 90mm) and included glass prefilters increase flow rates
  • Provided sterile and certified nonpyrogenic
Complete filter/storage system includes a polystyrene filter funnel joined by a color-coded polyethylene adapter to a removable PS storage bottle. The adjustable, angled hose connector simplifies vacuum-line attachment and ensures stability. Membrane material and pore size are printed on every unit for easy identification. Glass fiber prefilters are included.
Sterile caps for the easy-grip storage bottles are individually wrapped and designed to ensure a secure seal.
$165.24 - $487.54CAD / Case of 12
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Corning 431096 Disposable Vacuum Filtration Systems, 0.22um PES, 250 mL; 12/Cs 2953002 Mfr # 431096
COMPARE
Disposable Vacuum Filtration Systems, 0.22um PES, 250 mL; 12/Cs
$186.75CAD / Case of 12
Corning 430767 Disposable Vacuum Filtration Systems, 0.22um CA, 250 mL; 12/Cs 2953004 Mfr # 430767
COMPARE
Disposable Vacuum Filtration Systems, 0.22um CA, 250 mL; 12/Cs
$184.76CAD / Case of 12
Corning 430756 Disposable Vacuum Filtration Systems, 0.20um CN, 250 mL; 12/Cs 2953006 Mfr # 430756
COMPARE
Disposable Vacuum Filtration Systems, 0.20um CN, 250 mL; 12/Cs
$184.32CAD / Case of 12
Corning 431097 Disposable Vacuum Filtration Systems, 0.22um PES, 500 mL; 12/Cs 2953012 Mfr # 431097
COMPARE
Disposable Vacuum Filtration Systems, 0.22um PES, 500 mL; 12/Cs
$304.03CAD / Case of 12
Corning 430769 Disposable Vacuum Filtration Systems, 0.22um CA, 500 mL; 12/Cs 2953014 Mfr # 430769
COMPARE
Disposable Vacuum Filtration Systems, 0.22um CA, 500 mL; 12/Cs
$317.66CAD / Case of 12
Corning 430773 Disposable Vacuum Filtration Systems, 0.20um NYL, 500 mL; 12/Cs 2953018 Mfr # 430773
COMPARE
Disposable Vacuum Filtration Systems, 0.20um NYL, 500 mL; 12/Cs
$335.12CAD / Case of 12
Corning 431098 Disposable Vacuum Filtration Systems, 0.22um PES, 1000 mL; 12/Cs 2953022 Mfr # 431098
COMPARE
Disposable Vacuum Filtration Systems, 0.22um PES, 1000 mL; 12/Cs
$451.96CAD / Case of 12
Corning 430517 Disposable Vacuum Filtration Systems, 0.22um CA, 1000 mL; 12/Cs 2953024 Mfr # 430517
COMPARE
Disposable Vacuum Filtration Systems, 0.22um CA, 1000 mL; 12/Cs
$464.49CAD / Case of 12
Corning 430186 Disposable Vacuum Filtration Systems, 0.22um CN, 1000 mL; 12/Cs 2953026 Mfr # 430186
COMPARE
Disposable Vacuum Filtration Systems, 0.22um CN, 1000 mL; 12/Cs
$463.23CAD / Case of 12
Corning 430515 Disposable Vacuum Filtration Systems, 0.20um NYL, 1000 mL; 12/Cs 2953028 Mfr # 430515
COMPARE
Disposable Vacuum Filtration Systems, 0.20um NYL, 1000 mL; 12/Cs
$487.54CAD / Case of 12
Corning 431153 Disposable Vacuum Filtration Systems, 0.22um PES, 150 mL; 12/Cs 2953060 Mfr # 431153
COMPARE
Disposable Vacuum Filtration Systems, 0.22um PES, 150 mL; 12/Cs
$165.24CAD / Case of 12
Corning 431154 Disposable Vacuum Filtration System; 0.22um, CA Membrane, 150 mL 2953062 Mfr # 431154
COMPARE
Disposable Vacuum Filtration System; 0.22um, CA Membrane, 150 mL
$171.94CAD / Case of 12
Showing 1- 12 of 12