Cole-Parmer® Consistometer
Verify the consistency, viscosity, and flow rates of fluids
  • Meets military specification R-81294B for paint manufacturing
Use this consistometer to determine the consistency of viscous materials by measuring the distance that the material flows under its own weight in a given time interval. Consistometer checks against consistency, viscosity, and flow rate standards. Ideal for predetermining product formulas to standardize production lots.
Consistometer requires only 75 mL of sample for measurement. Engraved graduations in 0.5-cm divisions ensure accurate results; spring-loaded gate prevents premature sample flow. Stainless steel construction prevents corrosion.
$1,164.20 - $1,328.19CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Depth (in) Height (in) Width (in)
Availability Pricing
COMPARE
5.5 3.5 14
$1,164.20CAD / Each
COMPARE
3.5 4.125 16.25
$1,328.19CAD / Each