Compact Ultrasonic Test Sieve Cleaners
Clean your 8" or 200-mm diameter sieves safely and effectively
  • Cleaning program alternates between regular cleaning and a powerful pulse to produce excellent cleaning results
Simply brush sieve before inserting it into the 1.4-gallon (5.3-L) ultrasonic bath—vibrations up to 37 kHz dislodge particles from sieve frame and screen. Select the run time from 1 to 5 minutes in 1-minute intervals or from 5 to 30 minutes in 5-minute intervals. Degas mode is ideal for degassing HPLC solvents while the sample prep mode is for standard mixing, cleaning, and dissolving applications.
Unit features a stainless steel (SS) tank and outer casing. Overall dimensions are 13-3/4" (35 cm) H x 10-1/4" (26 cm) dia; tank measures 5-1/8" (13 cm) H x 9-7/16" (24 cm) dia.
$1,959.61 - $2,755.24CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Gal) Power (VAC) Description
Availability Pricing
Elma 112 0118-001 Compact Ultrasonic Test Sieve Cleaner, 115 VAC 5998735 Mfr # 112 0118-001
COMPARE
1.4 115 Compact Ultrasonic Test Sieve Cleaner, 115 VAC
$2,755.24CAD / Each
Elma 112 0119-001 Compact Ultrasonic Test Sieve Cleaner, 230 VAC 5998736 Mfr # 112 0119-001
COMPARE
1.4 230 Compact Ultrasonic Test Sieve Cleaner, 230 VAC
$1,959.61CAD / Each