Compact Ductless Air Filter System
Versatile system lets you customize to fit your application.
Capture and remove particles and volatile contaminants at their source. Compact air cleaner weighs only 30 lbs and fits virtually anywhere. Twin self-supporting extractor arms are made of durable polypropylene—they resist heat, chemicals, and abrasions. Ideal for two-station setups.
What's included: one 2-ply polyester ring-panel prefilter, one filter hold-down grid, one inlet grate, one inlet cover for two extractor arms, two extractor arms, two arm-hood attachments, and one arm-hole cap.
Complete your system with filters and accessories sold separately.
$2,755.24CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Height (in) Power (VAC) Description
Availability Pricing
Air Impurities Removal Systems Compact Ductless Air Cleaner, 350W, Black 3370339 Mfr # S987-2A
COMPARE
12.5 120 Compact Ductless Air Cleaner, 350W, Black
$2,755.24CAD / Each