Cole-Parmer® Universal Specific Gravity and Baume Dual-Scale Glass Hydrometer
This universal hydrometer is ideal for measuring both specific gravity and Baume degrees for liquids lighter and heavier than water. The temperature standard is 60° F (15.6° C). Hydrometer is plain form, individually seriealized, and includes a multilanguage statement of accuracy supplied by the manufacturer. Features wax parafin and steel shot ballast.
$66.30CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hydrometer Form Specific Gravity Range (Max) (%) Specific Gravity Range (Min) (%)
Availability Pricing
COMPARE
Plain 2 0.7
$66.30CAD / Each