Cole-Parmer® Universal Pipette Tips with Filter, Sterile
Protect against cross-contamination with tips free of human touch
  • Pure porous polyethylene filter protects against cross-contamination Durable polypropylene construction Noncytotoxic and nonhemolytic Sterilized and DNase, RNase, and pyrogen free
Cole-Parmer sterile pipette tips are constructed of high-grade polypropylene and a polyethylene filter which prevents liquids and aerosols from entering the body of your pipette. Tips are 99% Bacterial Aerosol Filtration Efficient (BFE), ASTM F21012, ISO 10993-5 MRM, minimum essential media elution, and GLP compliant.
ITEM(S) ON SALE
$136.10 - $229.48CAD / Case of 1000
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Type Sterile Tip Volume (mL)
Availability Pricing
COMPARE
Polyethylene Yes 0.01
$210.69CAD / Case of 960
COMPARE
Polyethylene Yes 0.1
$136.10CAD / Case of 960
$211.06CAD / Case of 960
COMPARE
Polyethylene Yes 0.2
$211.06CAD / Case of 960
COMPARE
Polyethylene Yes 1
$229.48CAD / Case of 1000
COMPARE
Polyethylene Yes 1.25
$148.00CAD / Case of 960
$229.48CAD / Case of 960