Cole-Parmer® Two-Channel Electronic Paperless Recorders
The most economical two-channel electronic recorder on the market!
  • View data in dual trend, dual digital, digital inversed, or split screened formats
  • User-friendly interface makes configuration and setup simple
  • Accepts voltage, current, thermocouples, RTDs, or TTL compatible pulse inputs
  • Adjustable chart speed from 0.75 inches/second to 1 inch/10 hours
This electronic digital paperless recorder offers two universally configurable isolated inputs for measuring DC voltage, DC current, thermocouples, RTDs as well as frequency and pulse inputs. Unit includes four internal alarm setpoints, two alarm relay outputs, one digital control input, and a maximum sample storage rate of 100 samples per second that can be set for both channels allowing you to capture short process signal anomalies.
The pushbutton menu interface allows simple set up and the bright LCD display is easy to read and can be configured to display real-time data in digital or trend formats. Store large amounts of data (up to 2 gigabytes) with the optional Compact Flash cards.
USB communication model (80816-02) provides access to data stored on Compact Flash cards or the ability to collect real-time data via USB port and included cable. Select the Ethernet communication model (80816-04) allowing you network access to data card files utilizing 10/100BaseT Ethernet via the RJ45 connector mounted on the back of the recorder.
The optional Navigator software makes reviewing, printing and exporting your data simple. Software will automatically start up Excel, convert historical data, load data into an organized spreadsheet along with displaying scales, date and time stamp information.
$1,245.01 - $1,547.06CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Output Type
Availability Pricing
COMPARE
1 SPST relay (0.5 A)
$1,245.01CAD / Each
COMPARE
1 SPST relay (0.5 A), USB
$1,517.59CAD / Each
COMPARE
1 SPST relay (0.5 A), Ethernet
$1,547.06CAD / Each