Cole-Parmer® Thermoplastic Elastomer (TPE) Tubing
Designed for fluid transfer and use in peristaltic pump applications
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
  • Excellent for use in peristaltic pump applications
  • Great alternative to silicone tubing
  • Does not contain PVC, DEHP, phthalates, or other plasticizers
  • Ultra-low extractables and leachables
  • Low gas and oxygen permeability
  • Low protein binding and nonpyrogenic material
  • Thermally weldable
  • USP Class VI, USP 661, and RoHS compliant and BPOG tested
The Cole-Parmer® Thermoplastic Elastomer (TPE) tubing is soft and flexible tubing that is durable enough to provide a long service life. This tubing is designed for use with peristaltic pumps. This TPE tubing does not contain PVC, DEHP, phthalates, or plasticizers, making them ideal for various pharmaceutical, food, and beverage applications.
24 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
COMPARE
1/32 3/32 1/32
$66.30CAD / Pkg of 1
COMPARE
3/32 5/32 1/32
$82.14CAD / Pkg of 1
COMPARE
3/32 7/32 1/16
$150.29CAD / Pkg of 1
COMPARE
5/8 7/8 1/8
$668.92CAD / Pkg of 1
COMPARE
1/16 1/8 1/32
$71.09CAD / Pkg of 1
COMPARE
3/4 7/8 1/16
$567.26CAD / Pkg of 1
COMPARE
3/4 1 1/8
$795.63CAD / Pkg of 1
COMPARE
1/16 3/16 1/16
$77.35CAD / Pkg of 1
COMPARE
1 1 1/4 1/8
$994.54CAD / Pkg of 1
COMPARE
1/8 3/16 1/32
$99.45CAD / Pkg of 1
COMPARE
1/8 1/4 1/16
$150.29CAD / Pkg of 1
COMPARE
3/16 1/4 1/32
$146.97CAD / Pkg of 1
COMPARE
3/16 5/16 1/16
$173.86CAD / Pkg of 1
COMPARE
3/16 3/8 3/32
$241.64CAD / Pkg of 1
COMPARE
3/16 7/16 1/8
$215.11CAD / Pkg of 1
COMPARE
1/4 3/8 1/16
$176.81CAD / Pkg of 1
COMPARE
1/4 7/16 3/32
$215.11CAD / Pkg of 1
COMPARE
1/4 1/2 1/8
$258.58CAD / Pkg of 1
COMPARE
5/16 7/16 1/16
$211.43CAD / Pkg of 1
COMPARE
5/16 1/2 3/32
$235.01CAD / Pkg of 1
Showing 1 - 20 of 24