Cole-Parmer® Submersible pH electrodes, 3-Ft
Reach further into solutions
  • Three-foot length for easier measuring in tanks
  • Sturdy design—permanently potted in sturdy ABS plastic housing
  • Submersible up to cable
Submersible electrodes operate from 32 to 176°>F (0 to 80°>C). Submerse up to the cable. Measure pH from 0 to 13. Electrodes are potted in a 3 ft L x 1-1/16'' diameter ABS sheath. Ideal for measurements in open containers such as tanks or troughs.
$484.38 - $565.78CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Basic pH/Temperature Meter
$484.38CAD / Each
COMPARE
Submersible 3' L pH Probe, SJ/ABS; BNC
$565.78CAD / Each
COMPARE
Submersible 3' L pH Probe, SJ/ABS/100Ohm RTD; Plug/BNC
$514.21CAD / Each
COMPARE
Submersible 3' L pH Probe, SJ/ABS/100Ohm RTD; Tinned/BNC
$565.78CAD / Each
COMPARE
Submersible 3' L pH Probe, DJ/ABS; BNC
$541.84CAD / Each
COMPARE
Submersible 3' L pH Probe, DJ/ABS/100Ohm RTD; Plug/BNC
$537.42CAD / Each
COMPARE
Submersible 3' L pH Probe, DJ/ABS/100Ohm RTD; Tinned/BNC
$554.73CAD / Each
COMPARE
Submersible 3' L pH Probe, DJ/ABS/1kOhm RTD; Spade/BNC
$514.95CAD / Each
Showing 1- 8 of 8