Cole-Parmer® Standard Series Manual Defrost Freezers
Precise and reliable freezing with advanced temperature control
  • Digital microprocessor controller with an adjustable range of -15°C to -25°C for accurate temperature management
  • Enhanced security with keyed door lock and magnetic gasket to ensure a tight, protective seal
  • Built-in safety features include audible and visual alarms, resettable Min/Max, and remote alarm contacts
  • Uses natural hydrocarbon refrigerants for a sustainable choice
Cole-Parmer® Standard Series Manual Defrost Freezers offer a significant upgrade in performance for the reliable storage of critical samples and supplies. Featuring a precise digital microprocessor temperature controller, these freezers provide consistent and accurate temperature management, with a manual defrost system to maintain efficiency. Safety and security are prioritized with remote alarm contacts, a keyed door lock, and an audible/visual alarm system. Built with environmentally friendly hydrocarbon refrigerants and backed by a comprehensive warranty, these freezers deliver dependable performance and peace of mind for laboratories and research facilities.
ITEM(S) ON SALE
$5,180.44 - $5,575.31CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Freezer Capacity (cu ft) Number of Doors Power (VAC)
Availability Pricing
COMPARE
14 1 120
$5,180.44CAD / Each
$6,475.55CAD / Each
COMPARE
17 1 120
$5,357.25CAD / Each
$6,696.56CAD / Each
COMPARE
20 1 120
$5,575.31CAD / Each
$6,969.13CAD / Each