Cole-Parmer® Square Wide-Mouth Plastic Bottles, PPCO
Fill and remove samples with ease
  • Leakproof polypropylene screw closures
  • Withstand temperatures to 275°F (135°C)
  • Autoclavable
These space-saving leakproof bottles can withstand temperatures to 275°F (135° C). The wide mouth is easy to fill, store, and transport dry chemicals and environmental samples. Square shape offers visible label surface so bottles are ideal as sample mailers.
$68.07 - $150.29CAD / Pkg of 12
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
175 38 Polypropylene Copolymer
$77.50CAD / Pkg of 12
COMPARE
250 43 Polypropylene Copolymer
$109.03CAD / Pkg of 12
COMPARE
500 53 Polypropylene Copolymer
$150.29CAD / Pkg of 12
COMPARE
1000 63 Polypropylene Copolymer
$120.08CAD / Pkg of 6
COMPARE
4000 100 Polypropylene Copolymer
$68.07CAD / Each