Cole-Parmer Spherical Aluminum and Glass Dewar Flasks
Spherical design lowers evaporation rate
Safely store and transport liquid nitrogen in these fully silvered and evacuated Dewars. Low liquid nitrogen evaporation—only 0.015 liter/hr on 5-liter vessel when covered. Narrow-neck glass vessel sits in a hammer-forged aluminum casing; foam rubber insulation protects against breakage. A heat-insulated cap and carrying handle are included.
$806.68 - $1,403.40CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Volume (Liters)
Availability Pricing
COMPARE
1
$806.68CAD / Each
COMPARE
3
$1,276.69CAD / Each
COMPARE
5
$1,403.40CAD / Each