Cole-Parmer® SD Laboratory Overhead Mixer
Variable-speed mixer designed for reliability and durability
  • Four-digit LED display indicates rpm, torque, time, and rotation direction Durable—no wear user serviceable parts Low/no maintenance brushless motor Adjustable tilt from 0 to 30 degrees allows proper impeller placement IP44 rated per IEC 60529 User-replaceable keyless chuck
Cole-Parmer SD laboratory mixer ensures accurate, dependable motor speed control with “soft-start” feature and smooth, gradual adjustment to the speed setting. Constant speed is maintained electronically via automatic compensation for changes in load torque demands at both high and low speeds. Built-in motor current compensation minimizes speed drift with changes in load. Motor current limiting protects the system in case of a locked rotor condition. Output circuit protection protects against inadvertent output shorting.
The mixer rotates at 50 to 2500 rpm with a maximum torque of 113 in-oz (80 N·cm) and has a through-shaft capability. Bracket hardware is preinstalled to enable installation of the adjustable tilt mounting bracket. The motor cable connector has a locking mechanism when the cable is connected to prevent unintended disconnection in the event of accidental force on the motor cable.
The precision digital controller is used to turn on and set the speed, timer, direction of rotation, and torque limits for the mixer motor. LED indicators identify the function being displayed, and a flashing LED and audible alarm indicate when limit conditions have been reached.
ITEM(S) ON SALE
$3,023.40 - $4,473.22CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Min Speed (rpm) Motor Type
Availability Pricing
COMPARE
2500 50 Brushed DC motor
$3,023.40CAD / Each
$3,779.25CAD / Each
COMPARE
2500 50 Brushed DC motor
$4,473.22CAD / Each
$5,591.52CAD / Each