Cole-Parmer® Sanitary Clamps, 304 Stainless Steel
Great for food, beverage, and dairy applications—clamps comply with FDA and 3A standards
  • Ideal for general-purpose, food, beverage, and pharmaceutical applications
  • Free technical application support available to assist with tubing selection
  • 304 stainless steel (SS) offers good chemical resistance
Use the sanitary clamps with sanitary fittings to create a leakproof connection. These easy-to-use clamps are made from 304 stainless steel to provides good chemical resistance.
$29.32 - $66.60CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Sanitary Clamp Size (Port 1) (InchShort) Description
Availability Pricing
COMPARE
Stainless Steel 1/2 to 3/4 Fitting, 304 Stainless Steel, Sanitary Clamp, 1/2" to 3/4"
$29.32CAD / Each
COMPARE
Stainless Steel 1 to 1-1/2 Fitting, 304 Stainless Steel, Sanitary Clamp, 1" to 1-1/2"
$34.33CAD / Each
COMPARE
Stainless Steel 2 Fitting, 304 Stainless Steel, Sanitary Clamp, 2"
$39.19CAD / Each
COMPARE
Stainless Steel 2-1/2 Fitting, 304 Stainless Steel, Sanitary Clamp, 2-1/2"
$46.12CAD / Each
COMPARE
Stainless Steel 3 Fitting, 304 Stainless Steel, Sanitary Clamp, 3"
$51.42CAD / Each
COMPARE
Stainless Steel 4 Fitting, 304 Stainless Steel, Sanitary Clamp, 4"
$66.60CAD / Each