Cole-Parmer® Pressure Sensors with Luer Fittings
Measure the static and dynamic pressure of gases and liquids accurately up to 75 psi
  • High-pressure piezo-resistive transducer provides highly accurate pressure sensing
  • Flow-through design reduces dead space and pressure drop
  • Save money with single-use pressure sensor
  • Manufactured in ISO 13485 certified facilities in ISO Class 7 cleanrooms
  • All fluid contact components meet USP Class VI and ISO 10993-5 requirements
  • Latex free and animal-derivative component free
  • Sterilize by gamma irradiation or ethylene oxide
The single-use pressure sensors feature a high-pressure piezo-resistive transducer to accurately measure static and dynamic pressures of gases and liquids up to 75 psi. Unobstructed, flow-through design minimizes dead space and pressure drops. Use in multi-stage depth filtration, TFF/crossflow filtration, chromatography processes, glass and disposable bioreactor pressure monitoring, and filling operations applications. Pressure sensors come with Luer fittings for quick, leak-free connections.
$144.98 - $160.23CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Sterile Wetted Materials
Availability Pricing
COMPARE
No Polysulfone
$144.98CAD / Each
COMPARE
Yes Polysulfone
$160.23CAD / Each
COMPARE
No Polycarbonate
$144.98CAD / Each