Cole-Parmer® Precision API Thermohydrometers, Combination form
  • NIST Traceable
  • Blue Spirit
  • Non-Mercury
  • Temperature of Standardization 60°F/60°F
  • Wax Paraffin and Steel Shot
  • Individually Serialized
These thermo-hydrometers feature non-mercury performance liquid and conform to the accuracy requirements for ASTM thermo-hydrometers. Each thermo-hydrometer is calibrated in a triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST. Individually packaged with a multi-language NIST Traceable Statement of Accuracy. Made in the USA.
$123.03 - $131.72CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
9/21 Degree API Combined Form Thermo-Hydrometer
$131.72CAD / Each
COMPARE
19/31 Degree API Combined Form Thermo-Hydrometer
$131.72CAD / Each
COMPARE
29/41 Degree API Combined Form Thermo-Hydrometer
$126.71CAD / Each
COMPARE
39/51 Degree API Combined Form Thermo-Hydrometer
$123.47CAD / Each
COMPARE
59/71 Degree API Combined Form Thermo-Hydrometer
$130.54CAD / Each
COMPARE
69/81 Degree API Combined Form Thermo-Hydrometer
$123.03CAD / Each