Cole-Parmer® Polystat® Programmable Stainless Steel Open-Top Bath
Handles liquid temperatures up to 150°C
  • Control panel swivels 180°, allowing viewing from almost any angle
  • Controller bridge removes easily for tank cleaning
  • Bath lid reduces evaporation and improves temperature stability
  • Complies with DIN 12876-1 Class I safety requirements for use with non-flammable liquids
These heated stainless steel circulating baths with programmable temperature controller handle temperatures up to 150°C. The controller features a 4 1/4” touch-screen display with on-screen icons and plain text prompts, allowing you to create and edit time and temperature programs quickly and easily. It stores 10 programs of up to 100 steps each. A variable-speed pressure/suction pump with open- and close-loop external circulation capability keeps the bath liquid at a uniform temperature.
The chemical-resistant top plate remains cool to the touch when operating at high temperatures, dampens operational sound, and is easily cleaned. User safety is enhanced by redundant overtemperature protection, low-liquid level float switch, and settable high and low temperature limits.
The controller supports 11 languages (English, French, German, Spanish, Chinese, Portuguese, Russian, Hindi, Arabic, Italian, and Korean) while bidirectional RS-232, addressable RS-485, and Ethernet communications facilitate remote control and data logging. USB ports enable you to log data directly to a flash drive or computer. A lidded opening is available for optional cooling coil.
$5,575.31CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Power (VAC) Reservoir Capacity (L)
Availability Pricing
COMPARE
150 120 10
$5,575.31CAD / Each
COMPARE
150 240 10
$5,575.31CAD / Each