Cole-Parmer® Polystat® Heated Coliform Bath
Streamline fecal coliform and E. coli testing by easily setting common test temperatures
  • Integral recirculating pump offers excellent temperature uniformity
  • Easily view readings on the backlit digital display from across the room to enhance monitoring
  • Hinged, gabled, see-through cover drains back into the bath when opened
Use this bath for standard method coliform testing, general bacteriological testing, incubation, sample tempering and general laboratory use. User-friendly controller can be easily set to 45.5, 44.5, 41.5, and 35.0°C, temperatures commonly used for coliform testing. Controller comes preset at 44.5°C which complies with EPA regulations for fecal coliform contamination testing as specified in "Standard Test Methods for the Examination of Water and Wastewater" (19th edition).
View actual and set point temperatures simultaneously on the large 3” x 2-1/8” LCD. Large bath opening makes it easy to place and remove samples. Gabled cover allows for taller vessels. Lidded port accommodates an optional cooling coil. Top deck remains cooler at high temperatures and is easily cleaned and disinfected.
$3,970.79CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Power (VAC) Reservoir Capacity (L)
Availability Pricing
COMPARE
135 120 28
$3,970.79CAD / Each
COMPARE
135 240 28
$3,970.79CAD / Each