Cole-Parmer® Platinum-Cured Silicone Tubing, 50 Shore A
Durable tubing offers long service life in peristaltic pump or pinch valve applications
  • Extruded to precise dimensional tolerances for accuracy
  • Tasteless/odorless tubing—helps maintain fluid integrity
  • Does not contain BPA or phthalates
  • Translucent tubing provides good clarity
  • Cleanroom packed and double bagged package
  • Sterilize by autoclave, gamma irradiation, EtO, and chemical sterilants
  • Complies with U.S. Pharmacopeia 43, National Formulary 38 (USP), General Chapter <88> Class VI, Biological reactivity Tests, In Vivo (2020)
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Produced with very high dimensional tolerances, the platinum-cured silicone tubing provides the accuracy and purity needed for critical biopharmaceutical applications. This soft yet durable tubing offers a long service life in peristaltic pump or pinch valve use.
Easily maintain the purity of the fluid—tubing does not contain BPA or phthalates that can leach out into the fluid. Tubing is manufactured, double-bagged, and sealed in an ISO Class 7 cleanroom. For an added level of particle control, a liner is placed between the double-bagged tubing package and the box.
$149.55 - $1,013.69CAD / Pkg of 1
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
COMPARE
1/16 3/16 1/16
$149.55CAD / Pkg of 1
COMPARE
1/8 1/4 1/16
$176.07CAD / Pkg of 1
COMPARE
3/16 5/16 1/16
$260.79CAD / Pkg of 1
COMPARE
1/4 1/2 1/8
$611.46CAD / Pkg of 1
COMPARE
3/8 5/8 1/8
$686.60CAD / Pkg of 1
COMPARE
3/16 7/16 1/8
$464.12CAD / Pkg of 1
COMPARE
1/4 3/8 1/16
$256.37CAD / Pkg of 1
COMPARE
1/4 7/16 3/32
$464.12CAD / Pkg of 1
COMPARE
1/4 3/4 1/8
$1,013.69CAD / Pkg of 1
Showing 1- 9 of 9