Cole-Parmer® Plastic Pasteur Pipettes
A safe alternative to glass pipettes
  • Manufactured from medical-grade polystyrene in an ISO Class 8 cleanroom
  • Gamma irradiated, and RNase, DNase, and pyrogen free
Disposable polystyrene (PS) Pasteur pipettes offer the aspirating precision of glass, with the safety and benefits of plastic. Ideal for use with aspirating bulbs or vacuum applications to remove liquid waste from flasks, dishes, plates, and tubes. The long flexible stem can be bent to draw liquid from narrow or small volume tubes. Supplied individually wrapped.
$129.36 - $133.34CAD / Pkg of 200
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Overall Length (InchShort)
Availability Pricing
COMPARE
5.7
$133.34CAD / Pkg of 200
COMPARE
9
$129.36CAD / Pkg of 200