Cole-Parmer® PES Sterile Chromatography Syringe Filter
  • Bidirectional flow with female luer lock on one side and a male luer slip on the other
  • Withstand pressure up to 100 psi (7 bar)
  • Batch test certificate included with each pack
PES Syringe Filters are naturally low protein binding and sample absorbing, and have a wide chemical compatibility for both aqueous and organic solvent filtration. Recommended for filtering critical biological sampling, tissue culture media, additives, serum and buffers. Maximum use temperature is 50°C (122°F).
$125.53 - $139.09CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Pore Size (µm) Sterile
Availability Pricing
COMPARE
25 0.2 Yes
$139.09CAD / Each
COMPARE
25 0.45 Yes
$125.53CAD / Each