Cole-Parmer® NPT(M) Threaded Plugs, PP
High-purity fittings deliver exceptional chemical resistance, durability, and compliance for critical fluid handling in food, bioprocess, and bioresearch applications
  • Injection molded from high-purity, semi-opaque fluoropolymer
  • Provides excellent chemical and corrosion resistance across harsh environments
  • Free from animal-derived materials, plasticizers, phthalates, and latex
  • Stable across a wide temperature range: -35 to 135°C (-31 to 275°F)
  • Supports ultrasonic welding for downstream processing flexibility
  • Sterilize by autoclave, gamma irradiation, or EtO
Engineered from a high-purity thermoplastic fluoropolymer, these PVDF fittings meet the rigorous demands of food, beverage, bioprocess, and bioresearch applications. This advanced material combines outstanding chemical resistance and corrosion protection with exceptional durability, maintaining performance across a wide temperature range from -35 to 135°C (-31 to 275°F).
$12.82 - $16.94CAD / Pkg of 10
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem Availability Pricing
COMPARE
$13.41CAD / Pkg of 10
COMPARE
$16.94CAD / Pkg of 10
COMPARE
$12.82CAD / Pkg of 10
COMPARE
$14.00CAD / Pkg of 10