Cole-Parmer® LED Fiber Optic Illuminators
Long-life LEDs save money and maintenance time
  • Stackable housing saves bench space
  • Interchangeable/replaceable fiber optics
  • Fan cooled for more powerHigher light spot uniformity
  • Fully adjustable and removable high quality fousing lenses
High-power white LEDs provide uniform color temperature, high light output, low heat, low power consumption, and extremely long life. Ideal for use in microscopy applications, inspection, forensics, machine vision, and assembly stations. Easily adjust the light output using the variable 10 to 100% intensity control. A solid state intensity control for more precise and repeatable control. Internal solid-state high performance DC regulated power supply optimizes consistent power to the LEDs. Built-in handle stays out of the way and allows for stacking of multiple units. Illuminator is available with a gooseneck light pipe or ring lights. Accepts all international power cords. Can switch from North America 115V to Euro 230V or India or UK by just changing out power cord.
$2,145.26 - $2,436.99CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Number of Light Pipe Power Cord Included Description
Availability Pricing
COMPARE
1 115 VAC, US plug LED Fiber Optic Illuminator, Single-Gooseneck Light Pipe; 115 VAC
$2,145.26CAD / Each
COMPARE
2 115 VAC, US plug LED Fiber Optic Illuminator, Dual-Gooseneck Light Pipe; 115 VAC
$2,436.99CAD / Each
COMPARE
2 230 VAC, European plug LED Fiber Optic Illuminator, Dual-Gooseneck Light Pipe; 230 VAC
$2,436.99CAD / Each