Cole-Parmer® Industrial Bimetal Thermometers
Read two scales simultaneously
  • Easy-to-read dual °F/°C scales provide enhanced flexibility
  • Anti-parallax dished dial for superior readability
  • Durable all-welded stainless steel construction for tough environments
  • Prevent dial fog, entry of moisture, interior corrosion, and coil freeze up with hermetically sealed case
  • Rugged NEMA 4X/IP66 protection
  • Recalibrate using the external reset screw
  • ±1% full-scale accuracy according to ASME B40.200 (B40.3 Grade A)
The Cole-Parmer® industrial bimetal thermometers feature NEMA 4X/IP66 protection for tough applications. Accuracy is ±1% full-scale when stem is immersed 2" in liquids or 4" in gases (immerse models with a 2.5" stem length 2" in gases). Stem length is measured from the top of the threads to the bottom of the stem tip. Stem diameter is 0.25” with a ½” NPT(M) back connection. Maximum overtemperature for thermometers with a max temperature of 250°F is 100% of span, and thermometers with a max temperature of 550°F is 50% of span. The ambient temperature range limits are –40 to 200°F (–40 to 93°C).
$106.08 - $144.39CAD / Each
29 variations of this product are available.
See All Product Options Below
29 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Dial Size (in) Max Temperature (° F) Stem Length (in)
Availability Pricing
COMPARE
3 250 2.5
$106.08CAD / Each
COMPARE
3 250 4
$106.08CAD / Each
COMPARE
3 250 6
$106.08CAD / Each
COMPARE
3 250 9
$106.08CAD / Each
COMPARE
3 250 12
$106.08CAD / Each
COMPARE
3 120 4
$106.08CAD / Each
COMPARE
3 120 6
$106.08CAD / Each
COMPARE
3 120 9
$106.08CAD / Each
COMPARE
3 120 12
$106.08CAD / Each
COMPARE
3 550 2.5
$106.08CAD / Each
COMPARE
3 550 4
$106.08CAD / Each
COMPARE
3 550 6
$106.08CAD / Each
COMPARE
3 550 9
$106.08CAD / Each
COMPARE
3 550 12
$106.08CAD / Each
COMPARE
5 250 2.5
$144.39CAD / Each
COMPARE
5 250 4
$144.39CAD / Each
COMPARE
5 250 6
$144.39CAD / Each
COMPARE
5 250 9
$144.39CAD / Each
COMPARE
5 250 12
$144.39CAD / Each
COMPARE
5 120 2.5
$144.39CAD / Each
Showing 1 - 20 of 29