Cole-Parmer® In-Line or Submersible pH Electrodes with Probe Guard
Reduce bulb damage with probe guard
  • Versatile design—threads on both ends for submersion or in-line use
  • Double-junction electrodes available; resist contamination from heavy metals, organics, or AgCI reactive compounds
In-line or submersible electrodes with probe guard operate from 23 to 176°>F (-5 to 80°>C) and have a PVC housing with a 3/4'' NPT(M) fitting on both ends. Epoxy body withstands pressures to 100 psi. Measure pH from 0 to 14. All electrodes measure 6-3/4''L x 1'' OD, with a 1-3/4'' insertion depth.
$297.62 - $422.86CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
pH electrode; single-junction, bnc connector
$332.99CAD / Each
COMPARE
Insertion/Submersion pH Probe; SJ/CPVC/100Ohm RTD; Plug/BNC
$397.82CAD / Each
COMPARE
Insertion/Submersion pH Probe; SJ/CPVC/100Ohm RTD; Tinned/BNC
$390.45CAD / Each
COMPARE
Insertion/Submersion pH Probe; SJ/CPVC/1khm RTD; Spade/BNC
$297.62CAD / Each
COMPARE
Insertion/Submersion pH Probe; SJ/CPVC/10khm RTD; Tinned ends
$346.25CAD / Each
COMPARE
Insertion/Submersion pH Probe; DJ/CPVC/10khm RTD; Tinned ends
$406.66CAD / Each
COMPARE
Insertion/Submersion pH Probe; DJ/CPVC/; /BNC
$347.72CAD / Each
COMPARE
Insertion/Submersion pH Probe; DJ/CPVC/100Ohm RTD; Plug/BNC
$422.86CAD / Each
COMPARE
Insertion/Submersion pH Probe; DJ/CPVC/100Ohm RTD; Tinned/BNC
$406.66CAD / Each
COMPARE
Insertion/Submersion pH Probe; DJ/CPVC/1kOhm RTD; Spade/BNC
$418.44CAD / Each
Showing 1- 10 of 10