Cole-Parmer® Heavy-Duty Carboys with Handles
Withstands temperature applications up to 135°C (275°F)
  • No leaks with polypropylene leakproof cap
  • Built-in shoulder handles for easy carrying
  • Shatterproof construction
  • Meets ISO 13485:2016 and ISO 9001:2015
  • USP Class VI certified
  • Autoclavable
Designed for large-volume liquid storage and dispensing, these heavy-duty carboys are compatible with most acids, bases, alcohols, aldehydes, esters, and aliphatic hydrocarbons. Constructed from high-quality laboratory-grade polypropylene resin materials that is shatterproof and conforms to USP Class VI standards. These carboys will save you money compared to stainless steel and fiberglass-reinforced containers.
$363.19 - $517.16CAD / Pkg of 1
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Material Outside Diameter (OD) (in)
Availability Pricing
COMPARE
10 Polypropylene(PP) 9 51/64
$363.19CAD / Pkg of 1
COMPARE
20 Polypropylene(PP) 11 1/5
$517.16CAD / Pkg of 1