Cole-Parmer® Graduated Polypropylene Beakers
Molded in graduations will not wear off
  • Graduated in mL and oz for easy conversion
  • Stacking ribs provide easy separation so beakers will not stick together
  • Accurate to ±2.5%
Featuring pour spouts, molded in graduations, and heat resistance from 5 to 248°F (-15 to 150°C) these polypropylene beakers are excellent for general use in the lab.
$62.47 - $76.98CAD / Pkg of 50
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Overall Height (in)
Availability Pricing
COMPARE
15 5 1 1/2
$63.36CAD / Pkg of 100
COMPARE
30 5 1 3/4
$69.62CAD / Pkg of 100
COMPARE
50 5 2
$65.20CAD / Pkg of 100
COMPARE
100 5 2
$62.47CAD / Pkg of 100
COMPARE
150 5 3 9/64
$70.28CAD / Pkg of 100
COMPARE
250 10 3 1/2
$68.88CAD / Pkg of 50
COMPARE
400 25 4 21/64
$76.98CAD / Pkg of 50
COMPARE
600 25 4 27/32
$63.36CAD / Pkg of 25
Showing 1- 8 of 8