Cole-Parmer® Graduated Cylinders, PMP
Transparent polymethylpentene (PMP) cylinders offer good chemical and heat resistance
  • Meet ISO Standard 6706
  • Autoclavable
Low-cost cylinders are calibrated “to contain” and feature molded graduations. Call for additional sizes.
$9.72 - $102.99CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
COMPARE
10 0.2 Polymethylpentene
$13.11CAD / Each
COMPARE
25 0.5 Polymethylpentene
$15.62CAD / Each
COMPARE
50 1 Polymethylpentene
$9.72CAD / Each
COMPARE
100 1 Polymethylpentene
$12.38CAD / Each
COMPARE
250 2 Polymethylpentene
$18.27CAD / Each
COMPARE
500 5 Polymethylpentene
$25.05CAD / Each
COMPARE
1000 10 Polymethylpentene
$49.80CAD / Each
COMPARE
2000 20 Polymethylpentene
$102.99CAD / Each
Showing 1- 8 of 8