Cole-Parmer® Fluorinated HDPE Wide-Mouth Plastic Bottles
Stop worrying about splitting and puncturing
  • mproved barrier for reduced solvent absorption and penetration
  • Leakproof FLPP closures
  • Stands up to many solvents
These fluorination bottles offer strong barrier properties and reduce solvent absorption and penetration on both inside and outside surfaces. Bottles include leakproof fluorinated polypropylene (FLPP) screw closures. Split- and puncture-resistant.
$64.09 - $169.81CAD / Pkg of 12
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
125 43 Fluorinated High Density Polyethylene (FLPE)
$81.48CAD / Pkg of 12
COMPARE
250 43 Fluorinated High Density Polyethylene (FLPE)
$122.44CAD / Pkg of 12
COMPARE
500 53 Fluorinated High Density Polyethylene (FLPE)
$169.81CAD / Pkg of 12
COMPARE
1000 63 Fluorinated High Density Polyethylene (FLPE)
$155.07CAD / Pkg of 6
COMPARE
2000 63 Fluorinated High Density Polyethylene (FLPE)
$64.09CAD / Each