Cole-Parmer® Fittings, Sanitary to Monobarb Adapter, Polypropylene
High-quality nonmetallic sanitary fittings for critical fluid applications
  • Meet US and EU standards for use in pharmaceutical process applications
  • ASME-BPE, USP Class VI, and FDA compliant
  • Sterilize by autoclave, EtO, or gamma irradiation
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
Easily transition from sanitary fitting to a barb connection using these adapters. These single-use polypropylene sanitary fittings are ideal for applications where metal is incompatible. Smooth inner surface and seamless construction prevent particle entrapment, eliminating potential bacterial growth. Sanitary fittings are great for cell culture media, aseptic fill and transfer, and sterile filling/emptying applications. Order a clamp and gasket to complete your system.
$42.29 - $59.30CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Connection Type Sanitary Clamp Size (Port 1) (in)
Availability Pricing
COMPARE
1/8 Sanitary x Hose barb 3/4" mini tri-clamp
$42.29CAD / Each
COMPARE
3/16 Sanitary x Hose barb 3/4" mini tri-clamp
$55.62CAD / Each
COMPARE
1/4 Sanitary x Hose barb 3/4" mini tri-clamp
$42.58CAD / Each
COMPARE
3/8 Sanitary x Hose barb 3/4" mini tri-clamp
$43.02CAD / Each
COMPARE
1/2 Sanitary x Hose barb 3/4" mini tri-clamp
$43.47CAD / Each
COMPARE
1/4 Sanitary x Hose barb 1-1/2" tri-clamp
$48.25CAD / Each
COMPARE
3/8 Sanitary x Hose barb 1-1/2" tri-clamp
$52.31CAD / Each
COMPARE
1/2 Sanitary x Hose barb 1-1/2" tri-clamp
$55.99CAD / Each
COMPARE
3/4 Sanitary x Hose barb 1-1/2" tri-clamp
$58.94CAD / Each
COMPARE
1 Sanitary x Hose barb 1-1/2" tri-clamp
$59.30CAD / Each
Showing 1- 10 of 10