Cole-Parmer® FEP Tubing
Chemically inert, thermal-resistant rigid tubing with optical clarity
  • Made of 100% virgin-grade fluorinated ethylene propylene (FEP)
  • Excellent chemical and thermal resistance
  • Great optical clarity—easily view your sample in use
  • Sterilize by autoclave, EtO, and chemical sterilants
  • Complies with FDA 21 CFR 177.1550, USP Class VI, NSF 51, NSF 61, NSF 372, and ASTM D2116
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Use the Cole-Parmer FEP fluoropolymer tubing in pharmaceutical, food, and laboratory applications. This chemically inert tubing offers excellent resistance to a broad range of solvents, bases, and strong acids and exhibits high thermal stability.
The nonstick property and low permeability help reduce the absorption/adsorption of fluids, minimizing the risk of fluid contamination. Optically clear tubing makes it easy to view your fluid for sight gauge and flow applications.
$125.24 - $659.71CAD / Pkg of 1
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
COMPARE
1/16 1/8 1/32
$130.25CAD / Pkg of 1
COMPARE
3/32 5/32 1/32
$181.96CAD / Pkg of 1
COMPARE
1/8 3/16 1/32
$219.90CAD / Pkg of 1
COMPARE
1/8 1/4 1/16
$376.08CAD / Pkg of 1
COMPARE
3/16 1/4 1/32
$234.64CAD / Pkg of 1
COMPARE
5/32 1/4 3/64
$322.67CAD / Pkg of 1
COMPARE
7/16 1/2 1/32
$509.42CAD / Pkg of 1
COMPARE
1/2 9/16 1/32
$659.71CAD / Pkg of 1
COMPARE
1/32 1/16 1/32
$125.24CAD / Pkg of 1
Showing 1- 9 of 9