Cole-Parmer® Ergonomic Cleanroom/ESD Vinyl Chairs

Class 100/ISO Class 5 ESD clean room chairs are static-dissipating and easy to clean
  • Static-dissipating, heavy-duty vinyl seat features molded polyurethane foam cushioning
  • Ergonomic seat tilts 8° forward, 3° backward, and locks in place
  • Rugged and durable weight capacity of 300 lbs.
These Class 100/ISO Class 5 ANSI/ESD-STM-12.1-2006 clean room chairs are made of molded polyurethane foam cushioning covered with easily cleaned antibacterial/antimicrobial vinyl and feature proper grounding, conductive, and dissipative static control materials.

Ergonomic seat tilts forward, backward, and locks in place for ergonomic posture support. The contoured waterfall seat cushion and an adjustable back rest with lumbar support for increased comfort during extended sitting durations.

Clean room chairs have been tested to Federal Standard 209E and ISO 14644-1 (Class 100/ISO Class 5) and ANSI/ESD-STM-12.1-2006 standards.

All chairs feature a smooth pneumatic lift. Choose from desk or bench height chairs with pneumatic adjustment. Chairs are also available in black or blue colors, with or without adjustable arm rest and foot ring. Choose ESD casters or ESD glides, both of which are nonmarring and suitable for hard or carpeted floors.
$1,010.75 - $1,327.52CAD / Each

12 variations of this product are available.

See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Material Description
Availability Pricing
COMPARE
Black Vinyl Clean Room ESD Ergonomic Chair, Vinyl, Desk Height, Black, No Arms, Chrome Casters
$1,010.75CAD / Each
COMPARE
Black Vinyl Clean Room ESD Ergonomic Chair, Vinyl, Desk Height, Black, Adjustable Arms, Chrome Casters
$1,217.02CAD / Each
COMPARE
Blue Vinyl Clean Room ESD Ergonomic Chair, Vinyl, Desk Height, Blue, Adjustable Arms, Chrome Casters
$1,217.02CAD / Each
COMPARE
Black Vinyl Clean Room ESD Ergonomic Chair, Vinyl, Desk Height, Black, Adjustable Arms, Gliders
$1,217.02CAD / Each
COMPARE
Blue Vinyl Clean Room ESD Ergonomic Chair, Vinyl, Desk Height, Blue, Adjustable Arms, Gliders
$1,217.02CAD / Each
COMPARE
Blue Vinyl Clean Room ESD Ergonomic Chair, Vinyl, Bench Height, Blue, No Arms, Chrome Casters, Chrome Foot Ring
$1,122.72CAD / Each
COMPARE
Blue Vinyl Clean Room ESD Ergonomic Chair, Vinyl, Bench Height, Blue, No Arms, Gliders, Chrome Foot Ring
$1,122.72CAD / Each
COMPARE
Black Vinyl Clean Room ESD Ergonomic Chair, Vinyl, Bench Height, Black, Adjustable Arms, Chrome Casters, Chrome Foot Ring
$1,327.52CAD / Each
COMPARE
Blue Vinyl Clean Room ESD Ergonomic Chair, Vinyl, Bench Height, Blue, Adjustable Arms, Chrome Casters, Chrome Foot Ring
$1,327.52CAD / Each
COMPARE
Black Vinyl Clean Room ESD Ergonomic Chair, Vinyl, Bench Height, Black, Adjustable Arms, Gliders, Chrome Foot Ring
$1,327.52CAD / Each
COMPARE
Blue Vinyl Clean Room ESD Ergonomic Chair, Vinyl, Bench Height, Blue, Adjustable Arms, Gliders, Chrome Foot Ring
$1,327.52CAD / Each
COMPARE
Black Vinyl Clean Room ESD Ergonomic Chair, Vinyl, Bench Height, Black, Adjustable Arms, Chrome Casters, No Foot Ring
$1,255.33CAD / Each
Showing 1- 12 of 12

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat