Cole-Parmer® Economy Narrow-Mouth Plastic Bottles HDPE
Chemical-resistant — stands up to most corrosives
  • Withstands freezer storage down to -100°C
  • Will not leak with PP screw caps
  • Meets USP Class VI requirements
These all-purpose bottles are popularly used for packaging, shipping, and storage because of their reliable durability. Rigid, translucent, and chemical-resistant, they can be used with most corrosives. All have leakproof polypropylene screw caps.
$17.68 - $210.69CAD / Pkg of 24
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
30 20 mm High Density Polyethyene (HDPE)
$17.68CAD / Pkg of 12
COMPARE
30 20 mm High Density Polyethyene (HDPE)
$88.85CAD / Pkg of 72
COMPARE
60 20 mm High Density Polyethyene (HDPE)
$19.15CAD / Pkg of 12
COMPARE
60 20 mm High Density Polyethyene (HDPE)
$96.95CAD / Pkg of 72
COMPARE
125 24 mm High Density Polyethyene (HDPE)
$24.16CAD / Pkg of 12
COMPARE
125 24 mm High Density Polyethyene (HDPE)
$125.09CAD / Pkg of 72
COMPARE
250 24 mm High Density Polyethyene (HDPE)
$48.18CAD / Pkg of 12
COMPARE
250 24 mm High Density Polyethyene (HDPE)
$210.69CAD / Pkg of 72
COMPARE
500 38 mm High Density Polyethyene (HDPE)
$53.04CAD / Pkg of 12
COMPARE
500 38 mm High Density Polyethyene (HDPE)
$186.38CAD / Pkg of 48
COMPARE
1000 38 mm High Density Polyethyene (HDPE)
$62.18CAD / Pkg of 6
COMPARE
1000 38 mm High Density Polyethyene (HDPE)
$180.12CAD / Pkg of 24
Showing 1- 12 of 12