Cole-Parmer® DRNase Free™ Surface Decontaminant
Instantly eliminate DNase and RNase from labware and surfaces
  • Works on glass, plastic, stainless steel, and benchtops
  • Replace carcinogenic chemicals, like DEPC
  • Phosphate free and biodegradable
The DRNase Free reagent surface decontaminant is ideal for items and surfaces used in PCR that cannot be autoclaved. Just spray and rinse. This nontoxic formula doesn’t leave a residue, so it won’t interfere with gel polymerization or enzymatic reactions.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Applications Format Volume (Liters)
Availability Pricing
COMPARE
Glass, plastic, stainless steel, and benchtops Liquid spray 0.95
Call For Price
COMPARE
Glass, plastic, stainless steel, and benchtops Liquid spray 3.8