Cole-Parmer® DC-400 Standard Desiccator Cabinets
Safely store humidity sensitive products
  • Easily view your samples through acrylic viewing panels
  • Durable, rugged construction— aluminum frame and stainless steel (SS) shelves
  • SS shelves with holes ensure uniform humidity throughout chamber
Desiccators provide dust-free storage for humidity sensitive products such as optical equipment, electronic parts, PCB boards, and reagents. Place a desiccant material in the cabinets to keep the environment dry. Once the desiccant is saturated, simply replace or regenerate the desiccant for reuse. A lock and key let you securely lock the cabinet to prevent unauthorized access.
$1,174.29 - $1,547.06CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Chamber Volume (Liters) Exterior Height (InchShort)
Availability Pricing
COMPARE
50 17
$1,174.29CAD / Each
COMPARE
70 22.9
$1,547.06CAD / Each