Cole-Parmer® Compost Bimetal Thermometers
Long stem designed for soil measurements
  • Hermetically sealed case to prevent entry of moisture, interior corrosion and coil freeze up
  • Anti-parallax dial for precise readings of both scales
  • Silicone dampened coil for reduction of pointer flutter on high-vibration application and provides improved response times
  • External adjustment
  • 304 stainless steel case and stem
From a 12" to a 48" long stem, these thermometers make compost and soil measurements easy.
$146.90 - $426.18CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Stem Length (in)
Availability Pricing
COMPARE
93 -20 12
$146.90CAD / Each
COMPARE
93 -20 24
$306.47CAD / Each
COMPARE
93 -20 36
$352.88CAD / Each
COMPARE
93 -20 48
$426.18CAD / Each