Cole-Parmer® Cleanroom Panel-Mount Female Luer to Hose Barb Fittings, Straight Adapter, Nylon
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Forms a leak-free luer connection with a quick simple turn
  • Single barb design tightly “grips” the tubing
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Quickly and easily transition from soft tubing to a luer connection using these adapters. Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. A single barb on one end of the fitting creates a tight seal to keep the tubing in place.
$11.64 - $22.69CAD / Pkg of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
COMPARE
3/32 Nylon
$11.64CAD / Pkg of 10
COMPARE
1/8 Nylon
$11.64CAD / Pkg of 10
COMPARE
1/8 Nylon
$17.83CAD / Pkg of 10
COMPARE
3/32 Nylon
$22.69CAD / Pkg of 10
COMPARE
1/16 Nylon
$22.69CAD / Pkg of 10
COMPARE
1/8 Nylon
$22.69CAD / Pkg of 10