Cole-Parmer® Cleanroom Large-Bore Female Luer Plugs, Straight Adapter
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Large-bore luer fittings offer higher flow rates compared to standard luers
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. Simply connect the plug to a large-bore male luer adapter to stop the fluid flow. Plugs are available in five different formulations—select one that best suits your application.
$33.74 - $61.88CAD / Pkg of 10
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Luer Connection Type (Port 1) Description
Availability Pricing
COMPARE
Polycarbonate Large Bore Female Luer Luer to Plug Fitting, Straight Adapter, CrystalVu™, Cleanroom Packed, Large Bore Female Luer; 10/Pk
$34.77CAD / Pkg of 10
COMPARE
Nylon Large Bore Female Luer Luer to Plug Fitting, Straight Adapter, White Nylon, Cleanroom Packed, Large Bore Female Luer; 10/Pk
$61.88CAD / Pkg of 10
COMPARE
PVDF Large Bore Female Luer Luer to Plug Fitting, Straight Adapter, Natural Kynar®, Cleanroom Packed, Large Bore Female Luer; 10/Pk
$60.70CAD / Pkg of 10
COMPARE
Nylon Large Bore Female Luer Luer to Plug Fitting, Straight Adapter, Natural Nylon, Cleanroom Packed, Large Bore Female Luer; 10/Pk
$33.74CAD / Pkg of 10
COMPARE
Polypropylene Large Bore Female Luer Luer to Plug Fitting, Straight Adapter, Animal-Derivative-Free Polypropylene, Cleanroom Packed, Large Bore Female Luer; 10/Pk
$34.04CAD / Pkg of 10