Cole-Parmer Chilling Incubator
Unique energy-efficient Peltier solid-state heating and chilling element eliminates the need for refrigerants and CFC
  • Peltier-based element heats and chills with precise control even at ambient—ideal for forced degradation of antibiotics, protein crystal growth, and applications that require ambient temperature control
  • Use the built-in RS-232 to connect to a 286 or better computer
  • Clear plastic inner door allows you to view your samples while maintaining chamber environment
  • Programmable digital display
$6,077.73 - $8,648.80CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Max Temperature (° C) Power (VAC)
Availability Pricing
COMPARE
1 70 115
$6,077.73CAD / Each
COMPARE
1 70 230
$6,151.40CAD / Each
COMPARE
2 70 115
$7,971.04CAD / Each
COMPARE
2 70 115
$8,648.80CAD / Each