Cole-Parmer® Calimat Infrared Calibrator
Easily calibrate your infrared calibrator
Simpy aim your IR thermometer at the black area and compare the temperature on the IR thermometer to the green reading on the temperature strip.
Instructions on calibration mat are provided in English and French.
Calibrate temperature range: 26 to 56°F (-3 to 13°C)
$8.84CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Temperature Accuracy Temperature Resolution (° F)
Availability Pricing
COMPARE
$8.84CAD / Each