Cole-Parmer® Angled, Long Handled Lab Sampling Spoons, HDPE
Angled bowl provides easy sample retrieval
  • Constructed of chemically inert, high-density polyethylene (HDPE)
  • Molded in ISO 8 (Class 100,000) clean room and individually bagged
  • Long handle helps to scoop from a distance without losing spoon inside vessel
  • Available sterile or non-sterile
Long handled lab spoons are ideal for sampling and dispensing powders, granules, or liquids from bottles, jars, and other confined areas while helping to avoid personal contact with sample materials. The angled head gathers materials from container corners more easily than straight spoons. Both the sterile and non-sterile spoons are individually bagged and can be used directly from the bag without the need for costly cleaning. Sterile spoons are gamma irradiated with a minimum dose of 25 kGy.
These food-grade, high-density polyethylene (HDPE) lab spoons are manufactured in an ISO 8 (Class 100,000) cleanroom. They conform to FDA CFR 177.1520, EU Regulations 10/2011, and EC Regulations 1935/2004, and are BSE/TSE free.
$484.75 - $612.19CAD / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Overall Length (cm) Sterile
Availability Pricing
COMPARE
10 36.2 No
$484.75CAD / Pkg of 100
COMPARE
10 36.2 Yes
$612.19CAD / Pkg of 100