Cole-Parmer® Amber Narrow-Mouth Plastic Bottles, HDPE
Meets USP requirements for storage of light-sensitive materials
  • Reduce UV light transmission
  • Withstands freezer storage down to -100°C
  • Will not leak with PP screw caps
  • Meets USP Class VI requirements
These amber bottles reduce UV light transmission and meet USP Class VI requirements for storage of light-sensitive materials. They are useful for packaging, shipping, and storage. Include leakproof polypropylene screw caps.
$40.22 - $124.94CAD / Pkg of 12
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Overall Height (in)
Availability Pricing
COMPARE
30 20 mm 2 29/64
$40.22CAD / Pkg of 12
COMPARE
60 20 mm 3 29/64
$50.10CAD / Pkg of 12
COMPARE
125 24 mm 3 57/64
$62.62CAD / Pkg of 12
COMPARE
250 24 mm 5 15/64
$85.01CAD / Pkg of 12
COMPARE
500 28 mm 6 11/16
$124.94CAD / Pkg of 12
COMPARE
1000 38 mm 8 1/2
$105.79CAD / Pkg of 6