Cole-Parmer® Alcohol Glass Hydrometers
Accurately measure proof of alcohol percent with lab-accredited Hydrometers
  • Used before and after fermentation to calculate the alcohol content as a percentage
  • Commonly used in winery, brewery, and food production operations
  • Calibrated in accredited labs against NIST traceable equipment
  • Metal ballast ensures the instruments floats upright
For determining proof of alcohol percent, all hydrometers are made according to the Bureau of Internal Revenue, US Treasure Department. The tolerances listed for these hydrometers are those outlined in Circular 555 of the National Bureau of Standards, Testing of Hydrometers. Each hydrometer is calibrated in a triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST. Individually packaged with a multi-language Statement of Accuracy indicating accuracy traceable to NIST.
$97.24 - $145.13CAD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Combined Alcohol Glass Hydrometer, 0-100% Tralle/0-200% Proof
$145.13CAD / Each
COMPARE
0/20 Percent Alcohol Proof – Ethyl Alcohol Hydrometer
$97.24CAD / Each
COMPARE
20/40 Percent Alcohol Proof – Ethyl Alcohol Hydrometer
$101.66CAD / Each
COMPARE
40/60 Percent Alcohol Proof – Ethyl Alcohol Hydrometer
$97.69CAD / Each
COMPARE
60/80 Percent Alcohol Proof – Ethyl Alcohol Hydrometer
$97.69CAD / Each
COMPARE
75/95 Percent Alcohol Proof – Ethyl Alcohol Hydrometer
$97.69CAD / Each
COMPARE
105/125 Percent Alcohol Proof – Ethyl Alcohol Hydrometer
$97.69CAD / Each
COMPARE
125/145 Percent Alcohol Proof – Ethyl Alcohol Hydrometer
$97.69CAD / Each
COMPARE
145/165 Percent Alcohol Proof – Ethyl Alcohol Hydrometer
$100.93CAD / Each
COMPARE
165/185 Percent Alcohol Proof – Ethyl Alcohol Hydrometer
$103.28CAD / Each
COMPARE
185/206 Percent Alcohol Proof – Ethyl Alcohol Hydrometer
$102.11CAD / Each
Showing 1- 11 of 11