Burkle TeleScoop Telescoping Open Container Sampler, Telescoping Rods
Versatile, secure, and easy to use
  • Attach different beakers and bottles to suit any sampling application
  • Telescoping rod sections feature end stops to prevent them from sliding apart
  • Sections do not rotate radially, providing a rigid handle for scooping viscous materials
  • User-friendly snap-in joint is practical and efficient in the field or on the floor
This modular sampling system allows you to build a sampler according to your application. Select a telescoping aluminum rod with the length you need, then choose one or more beakers and/or a bottle holder to attach to the rod. Angled and pendulum beakers are available in plastic or metal, and the bottle holder fits plastic, glass, or metal bottles.
$76.76 - $260.42CAD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (cm)
Availability Pricing
Burkle 5355-0120 Telescoping Rod, Aluminum, 65-120 cm; 2 Rods 0604209 Mfr # 5355-0120
COMPARE
120
$76.76CAD / Each
Burkle 5355-0180 Telescoping Rod, Aluminum, 70-180 cm; 4 Rods 0604210 Mfr # 5355-0180
COMPARE
180
$128.77CAD / Each
Burkle 5355-0250 Telescoping Rod, Aluminum, 125-250 cm; 2 Rods 0604211 Mfr # 5355-0250
COMPARE
250
$97.39CAD / Each
Burkle 5355-0280 Telescoping Rod, Aluminum, 95-280 cm; 4 Rods 0604212 Mfr # 5355-0280
COMPARE
280
$152.13CAD / Each
Burkle 5355-0300 Telescoping Rod, Aluminum, 115-300 cm; 3 Rods 0604213 Mfr # 5355-0300
COMPARE
300
$152.13CAD / Each
Burkle 5355-0450 Telescoping Rod, Aluminum, 165-450 cm; 3 Rods 0604214 Mfr # 5355-0450
COMPARE
450
$211.80CAD / Each
Burkle 5355-0600 Telescoping Rod, Aluminum, 65; 120 cm; 4 Rods 0604215 Mfr # 5355-0600
COMPARE
600
$260.42CAD / Each
Showing 1- 7 of 7