BrandTech® Dispensette® S Trace Analysis Bottletop Dispensers
Optimized for dispensing of ultra-pure chemicals
  • Improved flow technology primes quickly
  • Floating piston design reduces force needed to operate dispenser, even for large volumes
  • Discharge valve closes automatically when discharge tube is removed
  • Large viewport shows chemical location in discharge tube
  • Hinged screw cap swivels downward staying out of your way
Dispensers feature flowpaths made of EFTFE, FEP, PFA, PCTFE, PTFA, PTFE, and Al203 sapphire with a valve spring choice of either tantalum or platinum-iridium alloy. A special wiping seal cartridge ensures the integrity of your ultra-pure chemicals.
Dispensette S Trace Analysis Dispensers are recommended for high-purity applications and hydrofluoric acid. Choose between tantalum and platinum-iridium valve springs (use Pt-Ir for HF acid).
$2,335.32 - $2,401.63CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Dispense Volume (mL) Min Dispense Volume (mL) Description
Availability Pricing
BrandTech Dispensette® S Trace Analysis Bottletop Dispenser, Analog Adjustable, 1-10 mL, with Recirculation Valve, Platinum-Iridium Valve Spring 0791519 Mfr # 4640041
COMPARE
10 1 Trace Analysis Dispenser, 1-10 mL, Platinum-iridium Valve Spring, with Recirculation Valve
$2,401.63CAD / Each
BrandTech Dispensette® S Trace Analysis Bottletop Dispenser, Analog Adjustable, 1-10 mL, with Standard Valve, Platinum-Iridium Valve Spring 0791549 Mfr # 4640040
COMPARE
10 1 Trace Analysis Dispenser, 1-10 mL, Platinum-iridium Valve Spring, with Standard Valve
$2,335.32CAD / Each