Bottle and Cylinder Brushes
Ideal for cleaning bottles, cylinders, and large test tubes. Full radial end tip promotes effective cleaning and reduces scratching of glass and plasticware. Wire handle has hanging hook.
$268.16 - $411.44CAD / Pkg of 12
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Handle Length (in) Material
Availability Pricing
COMPARE
Black 7 Natural bristles, wire handle
$303.15CAD / Pkg of 12
COMPARE
Black 11 Nylon
$268.16CAD / Pkg of 12
COMPARE
Black 14 Nylon
$306.47CAD / Pkg of 12
COMPARE
Black 18 Nylon
$370.56CAD / Pkg of 12
COMPARE
Black 24 Nylon
$411.44CAD / Pkg of 12