Borosilicate Glass Tubing
Minimize expansion
  • Softening point acheived at 821°C (1509°F)
  • Working point is 1252°C (2885°F)
Manufactured of borosilicate glass, this standard wall tubing features great mechanical strength and low coefficient of expansion. Softening point is acheived at 821°C (1509°F) and working point is 1252°C (2885°F). A blast burner over gas can be used to bend smaller sizes and larger sizes must be heated over a blast burner using gas and oxygen. All sizes are cut 48'' in length.
$738.17 - $1,305.42CAD / Case of 380
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing OD (mm) Wall Thickness (mm)
Availability Pricing
COMPARE
4 0.8
$1,305.42CAD / Case of 380
COMPARE
5 0.8
$942.97CAD / Case of 256
COMPARE
7 1
$822.15CAD / Case of 120
COMPARE
9 1
$825.10CAD / Case of 164
COMPARE
11 1
$754.38CAD / Case of 132
COMPARE
12 1
$864.88CAD / Case of 120
COMPARE
14 1.2
$747.01CAD / Case of 96
COMPARE
15 1.2
$738.17CAD / Case of 80
COMPARE
16 1.2
$869.30CAD / Case of 49
COMPARE
19 1.2
$822.15CAD / Case of 62
COMPARE
20 1.2
$754.38CAD / Case of 58
COMPARE
25 1.5
$878.14CAD / Case of 37
COMPARE
28 1.5
$1,141.88CAD / Case of 33
COMPARE
38 2
$963.60CAD / Case of 18
Showing 1- 14 of 14