Binder Standard Series Solid.Line Incubators
Geared for research and quality assurance
  • APT.line™ preheating chamber technology
  • Runs on natural gravity convection
  • Adjustable exhaust for setting air change rates
  • Two-line LED display indicates actual chamber temperature and temperature set point—selectable °C or °F units
  • Controller offers timer with time delay feature and ramp function
  • Inner door features tempered safety glass
  • Class 3.1 integrated independent temperature safety device (DIN 12880) with visual alarm
Standard series of Solid.Line incubators are meant for easy everyday operation. Features such as digital temperature control, timer, and ramp function are perfect for a wide range of applications. An ergonomic door handle makes inserting and removing samples a breeze. Temperature range is ambient plus 5 to 70° C.
$2,616.74 - $3,555.29CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Max Temperature (° C) Power (VAC)
Availability Pricing
Binder BD-S 56 Solid.Line Gravity Convection Incubator, 2.2 cu ft; 120 VAC 3907324 Mfr # 9090-0017
COMPARE
2.2 70 120
$2,616.74CAD / Each
Binder BD-S 115 Solid.Line Gravity Convection Incubator, 4.2 cu ft; 120 VAC 3907326 Mfr # 9090-0023
COMPARE
4.2 70 120
$3,555.29CAD / Each