Binder FED Series Mechanical Convection Ovens with Enhanced Timer Functions
Designed for the requirements in volved in dryng or heating samples
  • Controller with LCD display and enhanced time functions
  • Electromechanical control of the exhaust air flap
  • Class 2 integrated independent adjustable temperature safety device (DIN 12880) with visual alarm
The FED series of ovens provide multiple capacities and are very adaptable to a variety of applications. LCD contoller makes it simple to adjust the fan speed, control the exhaust flap and adjust the enhanced timer functions.
With a temperature range up to 300°C these ovens maintain a high temperature accuracy thanks to APT.line™ technology. Other features include crome plated shelves, ethernet interface, USB port for data recording and an ergonomic handle design.
$5,053.73 - $10,229.75CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Power (VAC) Rise Time
Availability Pricing
Binder FED 115 Avantgarde Mechanical Convection Oven with Enhanced Timer Functions, 4.1 cu ft, 120 VAC 0501242 Mfr # 9010-0294
COMPARE
4.1 120 18 min to 150°C
$7,011.86CAD / Each
Binder FED 56 Avantgarde Mechanical Convection Oven with Enhanced Timer Functions, 2.1 cu ft, 230 VAC 0501243 Mfr # 9010-0295
COMPARE
2.1 230 15 min to 150°C
$5,053.73CAD / Each
Binder FED 56 Avantgarde Mechanical Convection Oven with Enhanced Timer Functions, 2.1 cu ft, 120 VAC 0501244 Mfr # 9010-0296
COMPARE
2.1 120 15 min to 150°C
$5,265.90CAD / Each
Binder FED 260 Avantgarde Mechanical Convection Oven with Enhanced Timer Functions, 9.1 cu ft, 240 VAC 0501267 Mfr # 9010-0300
COMPARE
9.1 240 19 min to 150°C
$10,229.75CAD / Each