Arsenic Test Kit
Measure arsenic levels in drinking water in only 12 minutes
  • Accurate, reliable results down to 2 ppb
  • Included in the EPA's Environmental Technology Verification program
Quickly and safely analyze samples. Only 2 minutes required for preparation, and only 10 minutes for incubation with this easy-to-use test kit. Reagents are in powder form and were carefully selected to minimize hazards normally associated with conventional arsenic testing kits.
$384.92 - $409.60CAD / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Measurement Points Test Tests Per Kit
Availability Pricing
ITS 481396 Water Quality Test Kit , Arsenic, Standard Range , 100/Pk RK-99531-32 Mfr # 481396
COMPARE
0, 5, 10, 20, 40, 60, 100, 200, 300, and 500 ppb Arsenic, standard range 100
$384.92CAD / Pkg of 100
ITS 481297 Water Quality Test Kit , Arsenic, Low Range , 50/Pk RK-99531-34 Mfr # 481297
COMPARE
0, 2, 4, 6, 8, 10, 15, 20, 30, 40, 60, 100, 160, and 240 ppb Arsenic, low range 50
$409.60CAD / Pkg of 50